Assay Details
| Gene Symbol: sppl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045244.2 | NP_001038709.2 | 9 - 10 | 1102 | 140 | |
| XM_021478906.1 | XP_021334581.1 | 9 - 10 | 1097 | 140 | ||
| XM_021478911.1 | XP_021334586.1 | 9 - 10 | 1097 | 140 | ||
| GenBank mRNA | AY926457.1 | - | 9 - 10 | 1086 | 140 | |
| BC044512.1 | - | 9 - 10 | 1070 | 140 | ||
| BC116574.1 | - | 9 - 10 | 1087 | 140 | ||
| EH571140.1 | - | 5 - 6 | 560 | 140 | ||
More Information
Set Membership: |
|

sppl2
Genomic Map