Assay Details
| Gene Symbol: crb2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045299.1 | NP_001038764.1 | 5 - 6 | 1418 | 71 | |
| GenBank mRNA | BC162451.1 | - | 5 - 6 | 1400 | 71 | |
| CU638819.1 | - | 5 - 6 | 1238 | 71 | ||
| DQ314736.1 | - | 5 - 6 | 1418 | 71 | ||
More Information
Set Membership: |
|

crb2a
Genomic Map