Assay Details
| Gene Symbol: bmf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045473.1 | NP_001038938.1 | 1 - 2 | 327 | 62 | |
| XM_005160866.4 | XP_005160923.1 | 2 - 3 | 495 | 62 | ||
| XM_009294967.3 | XP_009293242.1 | 2 - 3 | 500 | 62 | ||
| GenBank mRNA | DQ860156.1 | - | 1 - 2 | 327 | 62 | |
More Information
Set Membership: |
|

bmf2
Genomic Map