Assay Details
| Gene Symbol: nprl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045847.2 | NP_001039312.1 | 6 - 7 | 741 | 146 | |
| GenBank mRNA | BC122356.1 | - | 6 - 7 | 745 | 146 | |
| BC152088.1 | - | 6 - 7 | 717 | 146 | ||
More Information
Set Membership: |
|

nprl2
Genomic Map