Assay Details
| Gene Symbol: ppp1r14ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045853.1 | NP_001039318.1 | 4 - 5 | 450 | 134 | |
| XM_005161356.4 | XP_005161413.1 | 4 - 5 | 507 | 134 | ||
| GenBank mRNA | BC122190.1 | - | 4 - 5 | 450 | 134 | |
More Information
Set Membership: |
|

ppp1r14ba
Genomic Map