Assay Details
| Gene Symbol: klhl24a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077340.1 | NP_001070808.1 | 3 - 4 | 1172 | 60 | |
| GenBank mRNA | BC124660.1 | - | 3 - 4 | 1172 | 60 | |
More Information
Set Membership: |
|

klhl24a
Genomic Map