Assay Details
| Gene Symbol: ankrd10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077377.1 | NP_001070845.1 | 5 - 6 | 943 | 93 | |
| XM_005167453.4 | XP_005167510.1 | 3 - 4 | 1148 | 93 | ||
| GenBank mRNA | BC095671.1 | - | 5 - 6 | 844 | 93 | |
| BC124657.1 | - | 5 - 6 | 943 | 93 | ||
More Information
Set Membership: |
|

ankrd10b
Genomic Map