Assay Details
| Gene Symbol: skap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080011.2 | NP_001073480.2 | 10 - 11 | 1106 | 70 | |
| XM_021480127.1 | XP_021335802.1 | 10 - 11 | 1151 | 70 | ||
| GenBank mRNA | BC129161.1 | - | 10 - 11 | 1106 | 70 | |
More Information
Set Membership: |
|

skap1
Genomic Map