Assay Details
| Gene Symbol: col27a1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080575.1 | NP_001074044.1 | 57 - 58 | 4329 | 127 | |
| XM_021476201.1 | XP_021331876.1 | 37 - 38 | 127 | |||
| GenBank mRNA | EF032484.1 | - | 57 - 58 | 4329 | 127 | |
More Information
Set Membership: |
|

col27a1b
Genomic Map