Assay Details
| Gene Symbol: ptpdc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080596.2 | NP_001074065.2 | 3 - 4 | 379 | 59 | |
| XM_021479641.1 | XP_021335316.1 | 3 - 4 | 416 | 59 | ||
| GenBank mRNA | BC129183.1 | - | 3 - 4 | 361 | 59 | |
| BC131875.1 | - | 3 - 4 | 361 | 59 | ||
More Information
Set Membership: |
|

ptpdc1a
Genomic Map