Assay Details
| Gene Symbol: ptpdc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080596.2 | NP_001074065.2 | 8 - 9 | 2028 | 109 | |
| GenBank mRNA | BC129183.1 | - | 8 - 9 | 2010 | 109 | |
| BC131875.1 | - | 8 - 9 | 2010 | 109 | ||
More Information
Set Membership: |
|

ptpdc1a
Genomic Map