Assay Details
| Gene Symbol: pcsk5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083829.1 | NP_001077298.1 | 17 - 18 | 2560 | 79 | |
| GenBank mRNA | BC162145.1 | - | 17 - 18 | 2418 | 79 | |
| DQ983233.1 | - | 17 - 18 | 2560 | 79 | ||
More Information
Set Membership: |
|

pcsk5b
Genomic Map