Assay Details
| Gene Symbol: pcsk5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083829.1 | NP_001077298.1 | 14 - 15 | 2173 | 67 | |
| GenBank mRNA | BC162145.1 | - | 14 - 15 | 2031 | 67 | |
| DQ983233.1 | - | 14 - 15 | 2173 | 67 | ||
More Information
Set Membership: |
|

pcsk5b
Genomic Map