Assay Details
| Gene Symbol: npm2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123007.3 | NP_001116479.2 | 8 - 9 | 586 | 137 | |
| GenBank mRNA | EU543215.2 | - | 8 - 9 | 588 | 137 | |
More Information
Set Membership: |
|

npm2b
Genomic Map