Assay Details
| Gene Symbol: vps35l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098181.1 | NP_001091651.1 | 29 - 30 | 2799 | 64 | |
| GenBank mRNA | BC139716.1 | - | 29 - 30 | 2799 | 64 | |
More Information
Set Membership: |
|

vps35l
Genomic Map