Assay Details
| Gene Symbol: ppp1r8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100952.1 | NP_001094422.1 | 6 - 7 | 685 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ppp1r8a| Gene Symbol: ppp1r8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100952.1 | NP_001094422.1 | 6 - 7 | 685 | 77 | |
Set Membership: |
|