Assay Details
| Gene Symbol: tmem79a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109727.1 | NP_001103197.1 | 5 - 6 | 987 | 83 | |
| XM_009293920.3 | XP_009292195.1 | 6 - 7 | 1080 | 83 | ||
| XM_009293922.3 | XP_009292197.1 | 5 - 6 | 1058 | 83 | ||
| XM_021468145.1 | XP_021323820.1 | 5 - 6 | 1055 | 83 | ||
| GenBank mRNA | BC152294.1 | - | 5 - 6 | 987 | 83 | |
More Information
Set Membership: |
|

tmem79a
Genomic Map