Assay Details
| Gene Symbol: plxna1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110010.1 | NP_001103480.1 | 12 - 13 | 2991 | 70 | |
| GenBank mRNA | AB262186.1 | - | 12 - 13 | 2991 | 70 | |
| FJ209038.1 | - | 4 - 5 | 482 | 70 | ||
More Information
Set Membership: |
|

plxna1b
Genomic Map