Assay Details
| Gene Symbol: plxna1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110010.1 | NP_001103480.1 | 4330 | 94 | ||
| GenBank mRNA | AB262186.1 | - | 4330 | 94 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

plxna1b| Gene Symbol: plxna1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110010.1 | NP_001103480.1 | 4330 | 94 | ||
| GenBank mRNA | AB262186.1 | - | 4330 | 94 | ||
Set Membership: |
|