Assay Details
| Gene Symbol: plxna1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110010.1 | NP_001103480.1 | 19 - 20 | 4640 | 75 | |
| GenBank mRNA | AB262186.1 | - | 19 - 20 | 4640 | 75 | |
More Information
Set Membership: |
|

plxna1b
Genomic Map