Assay Details
| Gene Symbol: plxna1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110010.1 | NP_001103480.1 | 10 - 11 | 2736 | 78 | |
| GenBank mRNA | AB262186.1 | - | 10 - 11 | 2736 | 78 | |
| FJ209038.1 | - | 2 - 3 | 227 | 78 | ||
More Information
Set Membership: |
|

plxna1b
Genomic Map