Assay Details
| Gene Symbol: plxna1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110010.1 | NP_001103480.1 | 11 - 12 | 2850 | 80 | |
| GenBank mRNA | AB262186.1 | - | 11 - 12 | 2850 | 80 | |
| FJ209038.1 | - | 3 - 4 | 341 | 80 | ||
More Information
Set Membership: |
|

plxna1b
Genomic Map