Assay Details
| Gene Symbol: cnot6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110028.1 | NP_001103498.1 | 6 - 7 | 690 | 79 | |
| GenBank mRNA | BC153317.1 | - | 6 - 7 | 690 | 79 | |
More Information
Set Membership: |
|

cnot6b
Genomic Map