Assay Details
| Gene Symbol: rnf34b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110035.1 | NP_001103505.1 | 3 - 4 | 435 | 72 | |
| XM_005162428.4 | XP_005162485.1 | 2 - 3 | 362 | 72 | ||
| XM_005162429.4 | XP_005162486.1 | 2 - 3 | 362 | 72 | ||
| GenBank mRNA | BC151965.1 | - | 3 - 4 | 435 | 72 | |
More Information
Set Membership: |
|

rnf34b
Genomic Map