Assay Details
| Gene Symbol: chst3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110376.2 | NP_001103846.1 | 331 | 141 | ||
| GenBank mRNA | BC154463.1 | - | 332 | 141 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

chst3b| Gene Symbol: chst3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110376.2 | NP_001103846.1 | 331 | 141 | ||
| GenBank mRNA | BC154463.1 | - | 332 | 141 | ||
Set Membership: |
|