Assay Details
| Gene Symbol: kiss1rb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110531.1 | NP_001104001.1 | 1 - 2 | 238 | 80 | |
| XM_005168656.4 | XP_005168713.1 | 2 - 3 | 837 | 80 | ||
| XM_009302032.3 | XP_009300307.1 | 2 - 3 | 837 | 80 | ||
| GenBank mRNA | EU047918.1 | - | 1 - 2 | 238 | 80 | |
| HQ222883.1 | - | 1 - 2 | 238 | 80 | ||
| HQ222885.1 | - | 1 - 2 | 238 | 80 | ||
More Information
Set Membership: |
|

kiss1rb
Genomic Map