Assay Details
| Gene Symbol: kiss1rb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110531.1 | NP_001104001.1 | 2 - 3 | 375 | 65 | |
| XM_005168656.4 | XP_005168713.1 | 3 - 4 | 974 | 65 | ||
| XM_009302032.3 | XP_009300307.1 | 3 - 4 | 974 | 65 | ||
| GenBank mRNA | EU047918.1 | - | 2 - 3 | 375 | 65 | |
| HQ222887.1 | - | 2 - 3 | 458 | 65 | ||
More Information
Set Membership: |
|

kiss1rb
Genomic Map