Assay Details
| Gene Symbol: rbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130920.2 | NP_570995.1 | 4 - 5 | 390 | 72 | |
| XM_021480010.1 | XP_021335685.1 | 4 - 5 | 414 | 72 | ||
| GenBank mRNA | AJ236884.1 | - | 4 - 5 | 379 | 72 | |
| BC075787.1 | - | 4 - 5 | 390 | 72 | ||
| EF373650.1 | - | 4 - 5 | 379 | 72 | ||
More Information
Set Membership: |
|

rbp4
Genomic Map