Assay Details
| Gene Symbol: ccl27a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131062.2 | NP_571137.2 | 1 - 2 | 92 | 66 | |
| GenBank mRNA | AF201450.1 | - | 1 - 2 | 79 | 66 | |
| EH456016.1 | - | 1 - 2 | 115 | 66 | ||
More Information
Set Membership: |
|

ccl27a
Genomic Map