Assay Details
| Gene Symbol: ube2ib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131351.2 | NP_571426.1 | 2 - 3 | 191 | 96 | |
| XM_005174194.4 | XP_005174251.1 | 2 - 3 | 224 | 96 | ||
| GenBank mRNA | AF128240.1 | - | 2 - 3 | 124 | 96 | |
| BC059506.1 | - | 2 - 3 | 191 | 96 | ||
More Information
Set Membership: |
|

ube2ib
Genomic Map