Assay Details
| Gene Symbol: atp1b1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131668.3 | NP_571743.3 | 4 - 5 | 662 | 133 | |
| GenBank mRNA | AF286375.1 | - | 4 - 5 | 638 | 133 | |
| BC066590.1 | - | 4 - 5 | 662 | 133 | ||
More Information
Set Membership: |
|

atp1b1a
Genomic Map