Assay Details
| Gene Symbol: atp1b1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131668.3 | NP_571743.3 | 5 - 6 | 746 | 80 | |
| GenBank mRNA | AF286375.1 | - | 5 - 6 | 722 | 80 | |
| BC045376.1 | - | 5 - 6 | 756 | 80 | ||
| BC066590.1 | - | 5 - 6 | 746 | 80 | ||
| BC164078.1 | - | 5 - 6 | 686 | 80 | ||
More Information
Set Membership: |
|

atp1b1a
Genomic Map