Assay Details
| Gene Symbol: gfra1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131731.1 | NP_571806.1 | 4 - 5 | 810 | 67 | |
| GenBank mRNA | AY436321.1 | - | 4 - 5 | 810 | 67 | |
More Information
Set Membership: |
|

gfra1b
Genomic Map