Assay Details
| Gene Symbol: cry3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB042250.1 | - | 1 - 2 | 160 | 70 | |
| BC095305.1 | - | 2 - 3 | 323 | 70 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cry3a| Gene Symbol: cry3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB042250.1 | - | 1 - 2 | 160 | 70 | |
| BC095305.1 | - | 2 - 3 | 323 | 70 | ||
Set Membership: |
|