Assay Details
| Gene Symbol: cry3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313822.1 | NP_001300751.1 | 5 - 6 | 785 | 122 | |
| NM_131791.2 | NP_571866.2 | 4 - 5 | 609 | 122 | ||
| XM_005166892.4 | XP_005166949.1 | 5 - 6 | 788 | 122 | ||
| GenBank mRNA | AB042250.1 | - | 4 - 5 | 594 | 122 | |
| BC095305.1 | - | 5 - 6 | 757 | 122 | ||
| EH447304.1 | - | 5 - 6 | 783 | 122 | ||
More Information
Set Membership: |
|

cry3a
Genomic Map