Assay Details
| Gene Symbol: cry3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313822.1 | NP_001300751.1 | 6 - 7 | 874 | 86 | |
| NM_131791.2 | NP_571866.2 | 5 - 6 | 698 | 86 | ||
| XM_005166892.4 | XP_005166949.1 | 6 - 7 | 877 | 86 | ||
| GenBank mRNA | AB042250.1 | - | 5 - 6 | 683 | 86 | |
| BC095305.1 | - | 6 - 7 | 846 | 86 | ||
More Information
Set Membership: |
|

cry3a
Genomic Map