Assay Details
| Gene Symbol: cry3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313822.1 | NP_001300751.1 | 10 - 11 | 1680 | 80 | |
| NM_131791.2 | NP_571866.2 | 9 - 10 | 1504 | 80 | ||
| XM_005166892.4 | XP_005166949.1 | 10 - 11 | 1683 | 80 | ||
| GenBank mRNA | AB042250.1 | - | 9 - 10 | 1489 | 80 | |
| BC095305.1 | - | 10 - 11 | 1652 | 80 | ||
More Information
Set Membership: |
|

cry3a
Genomic Map