Assay Details
| Gene Symbol: efnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131805.1 | NP_571880.1 | 1 - 2 | 114 | 59 | |
| GenBank mRNA | AF375224.1 | - | 1 - 2 | 114 | 59 | |
| BC163323.1 | - | 1 - 2 | 210 | 59 | ||
| BC163326.1 | - | 1 - 2 | 210 | 59 | ||
| CU638799.1 | - | 1 - 2 | 1010 | 59 | ||
More Information
Set Membership: |
|

efnb1
Genomic Map