Assay Details
| Gene Symbol: efnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131805.1 | NP_571880.1 | 2 - 3 | 400 | 104 | |
| GenBank mRNA | AF375224.1 | - | 2 - 3 | 400 | 104 | |
| BC163323.1 | - | 2 - 3 | 496 | 104 | ||
| BC163326.1 | - | 2 - 3 | 496 | 104 | ||
| CU638799.1 | - | 2 - 3 | 1296 | 104 | ||
More Information
Set Membership: |
|

efnb1
Genomic Map