Assay Details
| Gene Symbol: ube2ia | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131833.2 | NP_571908.1 | 2 - 3 | 229 | 75 | |
| GenBank mRNA | AF332623.1 | - | 1 - 2 | 71 | 75 | |
| BC058302.1 | - | 2 - 3 | 229 | 75 | ||
| BC066609.1 | - | 2 - 3 | 207 | 75 | ||
More Information
Set Membership: |
|

ube2ia
Genomic Map