Assay Details
| Gene Symbol: ube2ia | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131833.2 | NP_571908.1 | 4 - 5 | 379 | 86 | |
| XM_021469272.1 | XP_021324947.1 | 3 - 4 | 316 | 86 | ||
| GenBank mRNA | BC058302.1 | - | 4 - 5 | 379 | 86 | |
| BC066609.1 | - | 4 - 5 | 357 | 86 | ||
More Information
Set Membership: |
|

ube2ia
Genomic Map