Assay Details
| Gene Symbol: casp3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131877.3 | NP_571952.1 | 4 - 5 | 600 | 77 | |
| GenBank mRNA | AB047003.1 | - | 3 - 4 | 319 | 77 | |
| BC078310.1 | - | 4 - 5 | 604 | 77 | ||
| KX084794.1 | - | 3 - 4 | 319 | 77 | ||
| MG957994.1 | - | 3 - 4 | 319 | 77 | ||
More Information
Set Membership: |
|

casp3a
Genomic Map