Assay Details
| Gene Symbol: gpib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144764.2 | NP_658910.2 | 17 - 18 | 1642 | 102 | |
| GenBank mRNA | AJ306396.1 | - | 17 - 18 | 1645 | 102 | |
| BC121194.1 | - | 17 - 18 | 1586 | 102 | ||
| EV564297.1 | - | 2 - 3 | 115 | 102 | ||
More Information
Set Membership: |
|

gpib
Genomic Map