Assay Details
| Gene Symbol: crx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152940.1 | NP_694419.1 | 1 - 2 | 507 | 75 | |
| GenBank mRNA | AF503443.1 | - | 1 - 2 | 507 | 75 | |
| BC059653.1 | - | 1 - 2 | 492 | 75 | ||
More Information
Set Membership: |
|

crx
Genomic Map