Assay Details
| Gene Symbol: spns1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153663.2 | NP_705949.2 | 10 - 11 | 1420 | 120 | |
| XM_005163832.4 | XP_005163889.1 | 10 - 11 | 1434 | 120 | ||
| GenBank mRNA | AF465772.1 | - | 10 - 11 | 1376 | 120 | |
| BC048024.1 | - | 10 - 11 | 1375 | 120 | ||
| BC165625.1 | - | 9 - 10 | 1220 | 120 | ||
| CF999294.1 | - | 2 - 3 | 268 | 120 | ||
| CK027241.1 | - | 5 - 6 | 630 | 120 | ||
More Information
Set Membership: |
|

spns1
Genomic Map