Assay Details
| Gene Symbol: spns1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153663.2 | NP_705949.2 | 2 - 3 | 443 | 118 | |
| XM_005163832.4 | XP_005163889.1 | 2 - 3 | 457 | 118 | ||
| GenBank mRNA | AF465772.1 | - | 2 - 3 | 399 | 118 | |
| BC048024.1 | - | 2 - 3 | 398 | 118 | ||
| BC165625.1 | - | 1 - 2 | 243 | 118 | ||
| CK029141.1 | - | 2 - 3 | 403 | 118 | ||
| EH595239.1 | - | 2 - 3 | 478 | 118 | ||
More Information
Set Membership: |
|

spns1
Genomic Map