Assay Details
| Gene Symbol: spns1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153663.2 | NP_705949.2 | 4 - 5 | 648 | 69 | |
| XM_005163832.4 | XP_005163889.1 | 4 - 5 | 662 | 69 | ||
| GenBank mRNA | AF465772.1 | - | 4 - 5 | 604 | 69 | |
| BC048024.1 | - | 4 - 5 | 603 | 69 | ||
| BC165625.1 | - | 3 - 4 | 448 | 69 | ||
| CK029141.1 | - | 4 - 5 | 608 | 69 | ||
| EH585608.1 | - | 4 - 5 | 658 | 69 | ||
| EH595239.1 | - | 4 - 5 | 683 | 69 | ||
More Information
Set Membership: |
|

spns1
Genomic Map