Assay Details
| Gene Symbol: tpi1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153668.4 | NP_705954.2 | 3 - 4 | 441 | 70 | |
| GenBank mRNA | AF387819.1 | - | 3 - 4 | 403 | 70 | |
| BC053294.1 | - | 3 - 4 | 381 | 70 | ||
| BC152271.1 | - | 3 - 4 | 393 | 70 | ||
| EH586742.1 | - | 3 - 4 | 447 | 70 | ||
More Information
Set Membership: |
|

tpi1b
Genomic Map