Assay Details
| Gene Symbol: atp6v1e1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173254.1 | NP_775361.1 | 7 - 8 | 724 | 98 | |
| GenBank mRNA | AF506222.1 | - | 7 - 8 | 724 | 98 | |
| BC057254.1 | - | 7 - 8 | 726 | 98 | ||
| BC067557.1 | - | 7 - 8 | 733 | 98 | ||
More Information
Set Membership: |
|

atp6v1e1b
Genomic Map