Assay Details
| Gene Symbol: u2af1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177479.2 | NP_803432.1 | 2 - 3 | 133 | 79 | |
| GenBank mRNA | AF506202.1 | - | 2 - 3 | 133 | 79 | |
| BC098601.1 | - | 2 - 3 | 160 | 79 | ||
More Information
Set Membership: |
|

u2af1
Genomic Map